Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoGAP p190 Mouse anti-Human, Mouse, Rat, Monkey, Clone: D2D6, MilliporeSigma™

Catalog No. 05378MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-378-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-378-MI Supplier MilliporeSigma Supplier No. 05378

Mouse Monoclonal Antibody

Detect RhoGAP p190 using this Anti-RhoGAP p190 Antibody, clone D2D6 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoGAP p190
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone D2D6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl with 0.05% sodium azide. Frozen solution.
Gene Accession No. NM_004308
Gene Symbols ARHGAP1, RHOGAP, p50-RhoGAP, CDC42GA1, CDC42GAP, Cdc42GAP, p50rhoGAP, RHOGAP1, RhoGAP
Host Species Mouse
Immunogen GST-fusion protein to amino acids 180-610 of rat p190 RhoGAP.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.