Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOH, Mouse, Clone: 3D3, Abnova™

Catalog No. 89016213 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-213 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-213 Supplier Abnova Corporation Supplier No. H00000399M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RHOH.

The protein encoded by this gene is a member of the Ras superfamily of small GTPases. Expression of a chimeric transcript of LAZ3 and this gene has been reported as a result of the translocation t(3;4) in non-Hodgkin's lymphomas. This gene encodes a small G-like protein, and unlike most other small G proteins which are expressed ubiquitously, this gene is transcribed only in hemopoietic cells. [provided by RefSeq]

Sequence: MLSSIKCVLVGDSAVGKTSLLVRFTSETFPEAYKPTVYENTGVDVFMDGIQISLGLWDTAGNDAFRSIRPLSYQQADVVLMCYSVANHNSFLNLKNKWIGEIRSNLPCTPVLVVVTQTDQREMGPHRASCVNAMEGKKLAQDVRAKGYLECSALSNRGVQQVFECAVRTAVNQARRRNRRRLFSINECKIF

Specifications

Antigen RHOH
Applications ELISA
Classification Monoclonal
Clone 3D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant RHOH.
Formulation PBS with no preservative; pH 7.4
Gene RHOH
Gene Accession No. BC014261
Gene Alias ARHH/TTF
Gene Symbols RHOH
Host Species Mouse
Immunogen RHOH (AAH14261.1, 1 a.a. ∼ 191 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.