Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIAP-3 (XIAP) Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07753
Description
Anti-RIAP-3 (XIAP) Antibody detects level of RIAP-3 (XIAP) and has been published and validated for use in western blotting.
Specifications
RIAP-3 (XIAP) | |
Polyclonal | |
BIRC4; HILP; XLP2; ILP1; Xiap; MIHA; hILP; IAP3; API3; XIAP | |
GST fusion protein corresponding to full-length rat inhibitor of apoptosis 3 (RIAP-3), also know as X-linked inhibitor of apoptosis (XIAP) | |
200 μg | |
NM_001167.2 | |
−20°C for 24 months | |
IgG |
Western Blot | |
Unconjugated | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction