Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ric-8B/Synembryn-B, Polyclonal

Catalog No. ABS1614MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1614MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1614MI Supplier MilliporeSigma™ Supplier No. ABS1614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Ric-8B/Synembryn-B Clone: in Human, Mouse, Rabbit samples, and it is validated for Immunoprecipitation, Western Blotting

Synembryn-B (UniProt Q80XE1; also known as Protein Ric-8B, Resistance to inhibitors of cholinesterase 8B) is encoded by the Ric8b (also known as BC051080) gene (Gene ID 237422) in murine species. Ric-8A and Ric-8B stimulate nucleotide exchange of all Gα subunit classes by stabilizing the Gα nucleotide-free transition state. Ric-8A acts upon Gαi/q/13, while Ric-8B targets Gαs/olf. In addition to their function as guanine nucleotide exchange factors (GEFs), Ric proteins also act as molecular chaperones that directly participate in the posttranslational folding of nascent Gα subunits to promote G-protein biosynthesis and membrane association. Ric-8 knockdown reduces Gα steady-state abundances and overexpression of Ric-8 proteins enhances cellular Gα levels. In Ric-8A– or Ric-8B–delicient ES cells, newly produced Gα subunits are defective in the initial association with an internal membrane, resulting in a ∼10-fold faster turned over rate than in WT cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ric-8B/Synembryn-B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q80XE1
Gene Symbols Ric8b
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Ric-8B/Synembryn-B.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_898995
Test Specificity Expected to react with all murine and human spliced isoforms (UniProt Q80XE1, Q9NVN3).
Target Species Human, Mouse, Rabbit
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.