Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIG-I Mouse anti-Human, Clone: 1C3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF297
Description
This Anti-RIG-I, clone 1C3 Antibody is validated for use in Western Blotting and Immunoprecipitation and Immunocytochemistry for the detection of RIG-I.
Specifications
RIG-I | |
Monoclonal | |
Unconjugated | |
NP_055129 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 λ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1C3 | |
Purified mouse monoclonal IgG1λ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
DDX58 | |
Recombinant protein corresponding to human RIG-I. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction