Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIMKLB, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RIMKLB peptide using ARM Technology.
Supplier: Abnova Corporation H00057494K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RIMKLB | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RIMKLB | |
RIMKLB | |
A synthetic peptide of human RIMKLB | |
RUO | |
57494 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ribosomal modification protein rimK-like family member B | |
RIMKLB | |
FAM80B, KIAA1238 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction