Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

regulating synaptic membrane exocytosis 2, Mouse, Clone: 3C12, Abnova™

Catalog No. 89001510 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-510 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-510 Supplier Abnova Corporation Supplier No. H00009699M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RIMS2.

Sequence: LANADLEDSRISMLRMDRPSRQRSISERRAAMENQRSYSMERTREAQGPSSYAQRTTNHSPPTPRRSPLPIDRPDLRRTD

Specifications

Antigen regulating synaptic membrane exocytosis 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RIMS2.
Formulation ascites with no preservative
Gene RIMS2
Gene Accession No. NM_014677
Gene Alias DKFZp781A0653/KIAA0751/OBOE/RAB3IP3/RIM2
Gene Symbols RIMS2
Host Species Mouse
Immunogen RIMS2 (NP_055492, 181 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9699
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.