Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RING1 Mouse anti-Human, Mouse, Clone: 4D6, MilliporeSigma™

Catalog No. 051362MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-136-2MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-136-2MI Supplier MilliporeSigma Supplier No. 051362
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-RING1 Antibody, clone 4D6 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry to detect RING1 also known as Polycomb complex protein RING1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RING1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4D6
Conjugate Unconjugated
Formulation Purified mouse monoclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002922.2
Gene Symbols EC 6.3.2.- , OTTHUMP00000029416, RING1A , RNF1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to RING1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.