Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIP3 Mouse anti-Human, Rat, Clone: 6E6.2, MilliporeSigma™

Catalog No. MABC28MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC28MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC28MI Supplier MilliporeSigma Supplier No. MABC28
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-RIP3 Antibody, clone 6E6.2 is validated for use in western blot, immunohistochemistry for the detection of RIP3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIP3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6E6.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_006862
Gene Symbols RIPK3, RIP3
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human RIP3 at the N-terminus.
Purification Method Protein G Purified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.