Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-RIPK1, Polyclonal, Novus Biologicals™

Supplier: Novus Biologicals NBP145841
Specifications
RIPK1/RIP1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal | |
Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Q13546 | |
RIPK1 | |
Synthetic peptide surrounding amino acid 645 of human RIP. | |
0.1 mg | |
Apoptosis | |
8737 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction