Learn More
receptor-interacting serine-threonine kinase 2, Mouse, Clone: 7F5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RIPK2.
Supplier: Abnova Corporation H00008767M05
Description
This gene encodes a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. The encoded protein contains a C-terminal caspase activation and recruitment domain (CARD), and is a component of signaling complexes in both the innate and adaptive immune pathways. It is a potent activator of NF-kappaB and inducer of apoptosis in response to various stimuli. [provided by RefSeq
Sequence: LQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVIVQKLKDNKQMGLQPYPEILVVSRSPSLNLLQNKSMSpecifications
receptor-interacting serine-threonine kinase 2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC004553 | |
RIPK2 | |
RIPK2 (AAH04553, 431 a.a. ∼ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Apoptosis | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
7F5 | |
Mouse monoclonal antibody raised against a partial recombinant RIPK2. | |
RIPK2 | |
CARD3/CARDIAK/CCK/GIG30/RICK/RIP2 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
8767 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.