Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIT1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA125573
Description
RIT belongs to the RAS (HRAS; MIM 190020) subfamily of small GTPases (Hynds et al., 2003 [PubMed 12668729]).
PA1-25573 detects RIT1 from human samples.PA1-25573 has been successfully used in Western blot procedures.The PA1-25573 immunogen is His-tagged recombinant human Rit1.
Specifications
RIT1 | |
Polyclonal | |
Unconjugated | |
Q92963 | |
RIT1 | |
His-tagged recombinant human Rit1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1mg/mL | |
RIT1 | |
MGC125864, MGC125865, RIBB, RIT, ROC1 | |
Rabbit | |
IgG fraction | |
RUO | |
6016 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction