Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Rituximab (Anti-Idiotype) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $573.00
Specifications
Antigen | Rituximab |
---|---|
Clone | 2260A |
Dilution | Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA, Binding Inhibition 2.5 ug/mL |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9630SP
![]() |
R&D Systems
MAB9630SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9630100
![]() |
R&D Systems
MAB9630100 |
100 μg |
Each for $573.00
|
|
|||||
Description
Rituximab Monoclonal specifically detects Rituximab in Multi-Species samples. It is validated for Western Blot, Flow Cytometry, ELISA, Binding Inhibition.Specifications
Rituximab | |
Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA, Binding Inhibition 2.5 ug/mL | |
Monoclonal | |
Supernatant | |
Multi-species | |
B lymphocyte antigen CD20, B lymphocyte surface antigen B1, Bp35, CD20, LEU 16, LEU16, Membrane spanning 4 domains subfamily A member 1, MS4A1 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2260A | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Full-length Rituximab | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Rituximab in Western blots in direct ELISAs. 100% cross-reactivity with Tositumomab and Ibritumomab (95% or higher sequence identity with Rituximab) and no cross-reactivity with Cetuximab, Trastuzumab, Brentuximab, Gemtuzumab, Adalimumab, Solanezumab, Teprotumumab, and Aducanumab is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title