Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RLA-DR Mouse anti-Rabbit, Biotin, Clone: RDR34, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 407008
Description
SouthernBiotech offers anti-rabbit RLA-DR and RLA-DQ antibodies for for use in a variety of applications.
Specifications
RLA-DR | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
Rabbit leukemic B cell line | |
RUO | |
Rabbit | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation | |
RDR34 | |
Biotin | |
Mouse | |
0.5 mg | |
Primary | |
Store at 2-8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction