Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RMND5B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512399
Description
This antibody is predicted to react with mouse based on sequence homology.

Specifications
RMND5B | |
Polyclonal | |
RMND5B | |
Protein RMD5 homolog B; UNQ2508/PRO5996; RMND5B | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
64777 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q96G75 | |
RMND5B | |
KLH conjugated synthetic peptide between 347-375 amino acids from the C-terminal region of human RMND5B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction