Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RNA Poly II, Milli-Mark™, FITC, Clone: CTD4H8,

Catalog No. FCMAB331FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB331FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB331FMI Supplier MilliporeSigma™ Supplier No. FCMAB331F

Specifications

Antigen RNA Poly II
Applications Flow Cytometry
Classification Monoclonal
Clone CTD4H8
Conjugate FITC
Formulation Purified mouse monoclonal IgG1κ conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_000928
Host Species Mouse
Immunogen Peptide corresponding to the carboxyl-terminal domain of RNA polymerase II.
Purification Method Protein G Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.