Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II Mouse anti-Drosophila, Human, Kangaroo, Clone: ARNA-3, MilliporeSigma™

Catalog No. CBL221MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL221MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL221MI Supplier MilliporeSigma Supplier No. CBL221
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect RNA Polymerase II with Anti-RNA Polymerase II Antibody, clone ARNA-3 (Mouse Monoclonal Antibody), that has been shown to work in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNA Polymerase II
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone ARNA-3
Conjugate Unconjugated
Formulation Supplied as tissue culture supernatant concentrate containing 0.09% sodium azide.
Gene Accession No. NM_057088.2
Gene Symbols KRT3, CK3, K3, CK-3, Cytokeratin-3, Keratin-3
Host Species Mouse
Immunogen Purified RNA Polymerase II
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Insect
Content And Storage 2-8°C for up to 12 months.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.