Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA polymerase II Mouse anti-Human, Mouse, Rat, Yeast, Clone: CTD4H8, MilliporeSigma™

Catalog No. 05623ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-623-ZMI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-623-ZMI Supplier MilliporeSigma Supplier No. 05623Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RNA polymerase II Antibody, clone CTD4H8, Ascites Free is a highly specific mouse monoclonal antibody, that targets RNA Polymerase and has been tested in western blotting, immunocytochemistry and ChIP-seq.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNA polymerase II
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone CTD4H8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. (This product is Ascites Free.)
Gene Accession No. NP_000928
Gene Symbols POLR2A, POLR2
Host Species Mouse
Immunogen Linear peptide corresponding to human RNA polymerase II.
Purification Method Protein G purified
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Yeast
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.