Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA polymerase II subunit B1 (phospho CTD Ser-2) Rat anti-Mouse, Clone: 3E10, MilliporeSigma™

Catalog No. 041571MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-157-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-157-1MI Supplier MilliporeSigma Supplier No. 041571
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2) Antibody, clone 3E10 is a Rat monoclonal antibody for detection of RNA polymerase II subunit B1 (phospho CTD Ser-2) has been validated in western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNA polymerase II subunit B1 (phospho CTD Ser-2)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E10
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000928
Gene Symbols POLR2A, POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corrresponding to human RNA polymerase subunit B1 CTD phosphorylated at Ser2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.