Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA polymerase II subunit B1 (phospho-CTD Ser-7) Rat anti-Mouse, Human, Clone: 4E12, MilliporeSigma™

Catalog No. 041570IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-157-0IMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-157-0IMI Supplier MilliporeSigma Supplier No. 041570I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12 is a highly specific rat monoclonal antibody and that targets RNA Polymerase and has been tested in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNA polymerase II subunit B1 (phospho-CTD Ser-7)
Applications Western Blot
Classification Monoclonal
Clone 4E12
Conjugate Unconjugated
Formulation Rat monoclonal IgG1κ with 0.05% sodium azide.
Gene Accession No. NP_000928
Gene Symbols POLR2A, POLR2
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to human RNA polymerase II subunit B1 phosphorylated at Ser7 of the C-terminal domain (CTD).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.