Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASET2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABF293MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF293MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF293MI Supplier MilliporeSigma Supplier No. ABF293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-RNASET2 Antibody detects level of RNASET2 and has been published and validated for use in RNASET2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASET2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols RNASET2; RNASE6PL
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to the C-terminus of human RNASET2.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.