Learn More
Description
Members of the Rho GTPase family, such as RND1, regulate the organization of the actin cytoskeleton in response to extracellular growth factors (Nobes et al., 1998 [PubMed 9531558]).[supplied by OMIM
Specifications
Specifications
| Antigen | RND1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human RND1 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RND1 |
| Gene Accession No. | NM_014470 |
| Gene Alias | ARHS/FLJ42294/RHO6/RHOS |
| Gene Symbols | RND1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.