Learn More
Description
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-DNA and protein-protein interactions. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq
Specifications
Specifications
| Antigen | ring finger protein 138 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3G2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant RNF138. |
| Formulation | ascites with no preservative |
| Gene | RNF138 |
| Gene Accession No. | NM_016271 |
| Gene Alias | HSD-4/MGC8758/NARF/STRIN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.