Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ring finger protein 19A, Mouse, Clone: 4E4, Abnova™

Catalog No. 89001530 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-530 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-530 Supplier Abnova Corporation Supplier No. H00025897M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RNF19.

The protein encoded this gene contains two RING-finger motifs and an IBR (in between RING fingers) motif. This protein is an E3 ubiquintin ligase that is localized in Lewy bodies (LBs), a characteristic neuronal inclusion in Parkinson's disease (PD) brains. This protein interacts with UBE2L3/UBCH7 and UBE2E2/UBCH8, but not other ubiquitin-conjugating enzymes. This protein is found to bind and ubiquitylate synphilin 1 (SNCAIP), which is a interacting protein of alpha synuclein in neurons, and a major component of LB. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]

Sequence: MKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTE

Specifications

Antigen ring finger protein 19A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RNF19.
Formulation ascites with no preservative
Gene RNF19A
Gene Accession No. NM_015435
Gene Alias DKFZp566B1346/DORFIN/RNF19
Gene Symbols RNF19A
Host Species Mouse
Immunogen RNF19 (NP_056250, 739 a.a. ∼ 837 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 25897
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.