Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF20 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89007928 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-928 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-928 Supplier Abnova Corporation Supplier No. H00056254B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RNF20 protein.

The protein encoded by this gene shares similarity with BRE1 of S. cerevisiae. Yeast BRE1 is a ubiquitin ligase required for the ubiquitination of histone H2B and the methylation of histone H3. [provided by RefSeq

Sequence: MSGIGNKRAAGEPGTSMPPEKKAAVEDSGTTVETIKLGGVSSTEELDIRTLQTKNRKLAEMLDQRQAIEDELREHIEKLERRQATDDASLLIVNRYWSQFDENIRIILKRYDLEQGLGDLLTERKALVVPEPEPDSDSNQERKDDRERGEGQEPAFSFLATLASSSSEEMESQLQERVESSRRAVSQIVTVYDKLQEKVELLSRKLNSGDNLIVEEAVQELNSFLAQENMRLQELTDLLQEKHRTMSQEFSKLQSKVETAESRVSVLESMIDDLQWDIDKIRKREQRLNRHLAEVLERVNSKGYKVYGAGSSLYGGTITINARKFEEMNAELEENKELAQNRLCELEKLRQDFEEVTTQNEKLKVELRSAVEQVVKETPEYRCMQSQFSVLYNESLQLKAHLDEARTLLHGTRGTHQHQVELIERDEVSLHKKLRTEVIQLEDTLAQVRKEYEMLRIEFEQTLAANEQAGPINREMRHLISSLQNHNHQLKGEVLRYKRKLREAQSDLNKTRLRSGSALLQSQSSTEDPKDEPAELKPDSEDLSSQSSASKASQEDANEIKSKRDEEERERERREKEREREREREKEKEREREKQKLKESEKERDSAKDKEKGKHDDGRKKEAEIIKQLKIELKKAQESQKEMKLLLDMYRSAPKEQRDKVQLMAAEKKSKAELEDLRQRLKDLEDKEKKENKKMADEDALRKIRAVEEQIEYLQKKLAMAKQEEEALLSEMDVTGQAFEDMQEQNIRLMQQLREKDDANFKLMSERIKSNQIHKLLKEEKEELADQVLTLKTQVDAQLQVVRKLEEKEHLLQSNIGTGEKELGLRTQALEMNKRKAMEAAQLADDLKAQLELAQKKLHDFQDEIVENSVTKEKDMFNFKRAQEDISRLRRKLETTKKPDNVPKCDEILMEEIKDYKARLTCPCCNMRKKDAVLTKCFHVFCFECVKTRYDTRQRKCPKCNAAFGANDFHRIYIG

Specifications

Antigen RNF20
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RNF20 protein.
Formulation No additive
Gene RNF20
Gene Accession No. NM_019592.5
Gene Alias BRE1/BRE1A/FLJ11189/FLJ20382/KIAA2779/MGC129667/MGC129668/hBRE1
Gene Symbols RNF20
Host Species Mouse
Immunogen RNF20 (NP_062538.5, 1 a.a. ∼ 975 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 56254
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.