Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF25, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008076 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-076 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-076 Supplier Abnova Corporation Supplier No. H00064320B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RNF25 protein.

The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination. [provided by RefSeq

Sequence: MAASASAAAGEEDWVLPSEVEVLESIYLDELQVIKGNGRTSPWEIYITLHPATAEDQDSQYVCFTLVLQVPAEYPHEVPQISIRNPRGLSDEQIHTILQVLGHVAKAGLGTAMLYELIEKGKEILTDNNIPHGQCVICLYGFQEKEAFTKTPCYHYFHCHCLARYIQHMEQELKAQGQEQEQERQHATTKQKAVGVQCPVCREPLVYDLASLKAAPEPQQPMELYQPSAESLRQQEERKRLYQRQQERGGIIDLEAERNRYFISLQQPPAPAEPESAVDVSKGSQPPSTLAAELSTSPAVQSTLPPPLPVATQHICEKIPGTRSNQQRLGETQKAMLDPPKPSRGPWRQPERRHPKGGECHAPKGTRDTQELPPPEGPLKEPMDLKPEPHSQGVEGPPQEKGPGSWQGPPPRRTRDCVRWERSKGRTPGSSYPRLPRGQGAYRPGTRRESLGLESKDGS

Specifications

Antigen RNF25
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RNF25 protein.
Formulation No additive
Gene RNF25
Gene Accession No. NM_022453.2
Gene Alias AO7/FLJ13906
Gene Symbols RNF25
Host Species Mouse
Immunogen RNF25 (NP_071898.2, 1 a.a. ∼ 459 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64320
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.