Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RNF5, Polyclonal

Catalog No. ST150250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST150250UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST150250UG Supplier MilliporeSigma™ Supplier No. ST150250UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Specifically detects RNF5 Clone: in Human samples, and it is validated for Immunoprecipitation

Expressed in a wide variety of tissues, RNF5 functions as a ubiquitin ligase that can regulate cell motility by controlling the ubiquitination of paxillin, a focal adhesion phosphoprotein that is localized to the cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF5
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids from a central region of human RNF-5
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽20kDa RNF5 protein in HeLa cells.
Target Species Human
Content And Storage −20°C
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.