Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF6 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89006155 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-155 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-155 Supplier Abnova Corporation Supplier No. H00006049B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RNF6 protein.

The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed. [provided by RefSeq

Sequence: MNQSRSRSDGGSEETLPQDHNHHENERRWQQERLHREEAYYQFINELNDEDYRLMRDHNLLGTPGEITSEELQQRLDGVKEQLASQPDLRDGTNYRDSEVPRESSHEDSLLEWLNTFRRTGNATRSGQNGNQTWRAVSRTNPNNGEFRFSLEIHVNHENRGFEIHGEDYTDIPLSDSNRDHTANRQQRSTSPVARRTRSQTSVNFNGSSSNIPRTRLASRGQNPAEGSFSTLGRLRNGIGGAAGIPRANASRTNFSSHTNQSGGSELRQREGQRFGAAHVWENGARSNVTVRNTNQRLEPIRLRSTSNSRSRSPIQRQSGTVYHNSQRESRPVQQTTRRSVRRRGRTRVFLEQDRERERRGTAYTPFSNSRLVSRITVEEGEESSRSSTAVRRHPTITLDLQVRRIRPGENRDRDSIANRTRSRVGLAENTVTIESNSGGFRRTISRLERSGIRTYVSTITVPLRRISENELVEPSSVALRSILRQIMTGFGELSSLMEADSESELQRNGQHLPDMHSELSNLGTDNNRSQHREGSSQDRQAQGDSTEMHGENETTQPHTRNSDSRGGRQLRNPNNLVETGTLPILRLAHFFLLNESDDDDRIRGLTKEQIDNLSTRHYEHNSIDSELGKICSVCISDYVTGNKLRQLPCMHEFHIHCIDRWLSENCTCPICRQPVLGSNIANNG

Specifications

Antigen RNF6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RNF6 protein.
Formulation No additive
Gene RNF6
Gene Accession No. NM_005977.2
Gene Alias DKFZp686P0776
Gene Symbols RNF6
Host Species Mouse
Immunogen RNF6 (NP_005968.1, 1 a.a. ∼ 685 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6049
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.