Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RNF8, Clone: EPR6312(2), Rabbit Monoclonal,

Catalog No. MABE617MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE617MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE617MI Supplier MilliporeSigma™ Supplier No. MABE617

Specifications

Antigen RNF8
Applications Western Blot
Classification Monoclonal
Clone EPR6312(2)
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human RNF8.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) RNF8
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.