Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNMT (mRNA methyltransferase) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 061355MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-135-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-135-5MI Supplier MilliporeSigma Supplier No. 061355

Rabbit Polyclonal Antibody

Anti-RNMT (mRNA methyltransferase) Antibody is a rabbit polyclonal antibody for detection of RNMT also known as mRNA cap guanine-N7 methyltransferase, RG7MT1, hCMT1, hMet, hcm1p and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNMT (mRNA methyltransferase)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols RNMT
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human RNMT.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003790
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.