Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO2, Mouse, Clone: 4G5, Abnova™

Catalog No. 89123114 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-114 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-114 Supplier Abnova Corporation Supplier No. H00006092M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ROBO2.

This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: ATGDPLPVISWLKEGFTFPGRDPRATIQEQGTLQIKNLRISDTGTYTCVATSSSGETSWSAVLDVTESGATISKNYDLSDLPGPPSKPQVTDVTKNSVTL

Specifications

Antigen ROBO2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ROBO2.
Formulation ascites with no preservative
Gene ROBO2
Gene Accession No. NM_002942
Gene Alias KIAA1568/SAX3
Gene Symbols ROBO2
Host Species Mouse
Immunogen ROBO2 (NP_002933, 441 a.a. ∼ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6092
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.