Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK-II Mouse, Unlabeled, Clone: 21, BD

Catalog No. BDB610624 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610624 150 μg
BDB610623 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610624 Supplier BD Biosciences Supplier No. 610624
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Activity of the GTP-binding proteins is regulated by GAPs, that accelerate binding, and GTPases, that enhance the rate of GTP hydrolysis (Ras, Rho, cdc42Hs, and Rac). The isoforms of Rho, a small GTP-binding protein, regulate cellular processes such as the formation of the stress fibers, lamelipodia, and filopodia. ROKα (RhoA-binding kinase) is a Ser/Thr protein kinase that interects with the GTP-binding form of RhoA. Like RhoA, ROKα localizes at the cellular margins and colocalizes with acti filaments. The ROKα gene encodes a protein of 1302 amino acids with homology to the human myotonic kinase. Although ROKα binds to GTP-RhoA, it is not yet clear whether this interaction induces the kinase activity of ROKα.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ROCK-II
Applications Western Blot
Classification Monoclonal
Clone 21
Concentration 250μg/mL
Conjugate Unconjugated
Description ROKα
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat ROKα aa. 567-718
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.