Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROD1 regulator of differentiation 1 (S. pombe), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ROD1.
Supplier: Abnova Corporation H00009991A01
Description
ROD1 is a functional homolog of nrd1, an S. pombe RNA-binding protein that suppresses the onset of differentiation.[supplied by OMIM
Sequence: GSDELLSSGIINGPFTMNSSTPSTANGNDSKKFKRDRPPCSPSRVLHLRKIPCDVTEAEIISLGLPFGKVTNLLMLKGKSQAFLEMASEEAAVTMVNYYSpecifications
| ROD1 regulator of differentiation 1 (S. pombe) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ROD1. | |
| ROD1 | |
| DKFZp781I1117/PTBP3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 9991 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005156 | |
| ROD1 | |
| ROD1 (NP_005147, 16 a.a. ∼ 114 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction