Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROKα/ROCK-II Rabbit anti-Human, Mouse, Rat, Bovine, Clone: A9W4, MilliporeSigma™

Catalog No. 04841MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-841-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-841-MI Supplier MilliporeSigma Supplier No. 04841
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Detect ROKα/ROCK-II using this Anti-ROKα/ROCK-II Antibody, clone A9W4 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROKα/ROCK-II
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A9W4
Conjugate Unconjugated
Formulation Cultured supernatant containing 0.05% sodium azide.
Gene Accession No. NM_013022
Host Species Rabbit
Immunogen 6His-tagged fusion protein corresponding to residues 2-543 of rat ROKα/ROCK-II.
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) EC 2.7.11.1, MGC131603, MGC43611, OTTHUMP00000162514, P160ROCK, PRO0435
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.