Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RORgammat Mouse anti-Human, Alexa Fluor 647, Clone: Q21-559, BD

Catalog No. BDB563620 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563620 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563620 Supplier BD Biosciences Supplier No. 563620
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Q21-559 monoclonal antibody recognizes ROR gamma t (RORγt), an isoform of RORgamma (RORγ). RORγt is a DNA-binding transcription factor that is encoded by the RORC (RAR-related orphan receptor C) gene. It belongs to the ROR/RZR orphan nuclear receptor family. Two protein isoforms are produced from the RORC gene by alternative splicing: RORγγt is highly restricted to the thymus where it is expressed exclusively in immature CD4+CD8+ thymocytes and in peripheral Th17 and LTi cells. RORγt reportedly functions as an essential transcription factor for the development of secondary lymphoid tissues. It also plays an important regulatory role in thymopoiesis and effector T cell differentiation, especially for Th17 cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RORgammat
Applications Flow Cytometry
Classification Monoclonal
Clone Q21-559
Conjugate Alexa Fluor 647
Description RORgt; ROR gamma t; RORC; NR1F3; RORG; RZRG; TOR; RZR-GAMMA
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human RORgamma t Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.