Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RORgammat Mouse anti-Human, Alexa Fluor 488, Clone: Q21-559, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 563621

Catalog No. BDB563621


Only null left
Add to Cart

Description

Description

The Q21-559 monoclonal antibody recognizes ROR gamma t (RORγt), an isoform of ROR gamma (RORγ). RORγt is a DNA-binding transcription factor that is encoded by the RORC (RAR-related orphan receptor C) gene. It belongs to the ROR/RZR orphan nuclear receptor family. Two protein isoforms are produced from the RORC gene by alternative splicing: ROR γγt is highly restricted to the thymus where it is expressed exclusively in immature CD4+CD8+ thymocytes and in peripheral Th17 and LTi cells. ROR γt reportedly functions as an essential transcription factor for the development of secondary lymphoid tissues. It also plays an important regulatory role in thymopoiesis and effector T cell differentiation, especially for Th17 cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RORgammat
Monoclonal
Alexa Fluor 488
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human RORgamma t Recombinant Protein
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
Q21-559
RORgt; ROR gamma t; RORC; NR1F3; RORG; RZRG; TOR; RZR-GAMMA
Mouse
Affinity Purified
RUO
Human
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.