Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-ROR2 Clone: Nt2535-2835,

Catalog No. MABS1152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1152MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1152MI Supplier MilliporeSigma™ Supplier No. MABS1152
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects ROR2 Clone: Nt2535-2835 in Human, Mouse samples, and it is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting

Tyrosine-protein kinase transmembrane receptor ROR2 (EC 2.7.10.1; UniProt Q9Z138; also known as mRor2, Ntrkr2, Neurotrophic tyrosine kinase receptor-related 2) is encoded by the Ror2 gene (Gene ID 26564) in murine species. Ror2 is a receptor tyrosine kinase that binds Wnt5a and negatively regulates canonical Wnt signaling. Although in C. elegans and Xenopus Ror2 is reported to modulate Wnt signaling nonautonomously by preventing Wnt from stimulating adjacent cells, studies conducted in mammalian cells suggest that Ror2 cytoplasmic domain is crucial for its function and Ror2 inhibits Wnt signaling in a cell-autonomous manner. Ror2 and the closely related Ror1 play essential roles during development. Human ROR2 gene mutations have been associated with dominant Brachydactyly B and recessive Robinow syndrome. Ror2-null mice exhibit skeletal abnormalities similar to the human syndromes. In addition, these mice display heart and lung defects and die perinatally. Ror2 is initially translated with an N-terminal signal sequence (a.a. 1-33), the removal of which yields the mature receptor with an extracellular region (a.a. 34-403), a transmembrane segment (a.a. 404-424), followed by the cytoplasmic region (a.a. 425-944) that contains the kinase domain (a.a. 473-746), a Pro-rich domain (a.a. 784-857) sandwiched between two Ser/Thr-rich domains (a.a. 753-782 and a.a. 859-882).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Nt2535-2835
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9Z138
Gene Symbols Ror2
Host Species Mouse
Immunogen Recombinant protein corresponding to the second Ser/Thr-rich domain of mouse mRor2.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone Nt 2535-2835 is specific to the mouse and human ROR2, but does not detect mouse ROR1. Clone Nt 2535-2835 exclusively detects ROR2 protein in paraffin-embedded tissues (Mikels, A., et al. (2009). J. Biol. Chem. 284(44):30167-30176).
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.