Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RORgt Mouse, Unlabeled, Clone: O28-835, BD

Catalog No. BDB562197 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562197 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562197 Supplier BD Biosciences Supplier No. 562197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The O28-835 monoclonal antibody recognizes ROR-gamma ( RORγ) and its isoform, RORγt. RORγ(∽58 kDa calculated molecular weight) is a DNA-binding transcription factor that belongs to the ROR/RZR orphan nuclear receptor family. It is expressed in different tissues including skeletal muscle, kidney, liver and adipose tissues. RORγt ( ∽56 kDa calculated molecular weight ) is a RORγisoform that is expressed exclusively by lymphoid cells. It is expressed by various lymphoid cell types including CD4+CD8+ thymocytes, peripheral CD4+ Th17 and CD8+ Tc17 cells and lymphoid tissue inducer (Lti) cells. RORγt plays essential roles in thymopoiesis, T cell homeostasis, differentiation of effector T lymphocytes and the development of secondary lymphoid tissues including lymph nodes and Peyer’s patches.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RORgt
Applications Western Blot
Classification Monoclonal
Clone O28-835
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Rorc; Rorg; RORgamma; Rorγ; RORgt; RORgammat; RORγt; TOR; Thor; Nr1f3; ROR
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human RORγt Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.