Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rotavirus Goat anti-Human, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AB1129
Description
This Anti-Rotavirus Antibody is validated for use in ELISA, immunofluoresence, western blot, immunohistochemistry for the detection of Rotavirus.
Specifications
Rotavirus | |
Polyclonal | |
Purified immunoglobulin. In PBS (0.01 M, pH 7.2) with 0.1% sodium azide | |
Bovine (Nebraska Calf Diarrhea Virus) | |
Primary | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Goat | |
1 Unit | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction