Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP9, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89202839 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-202-839 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-202-839 Supplier Abnova Corporation Supplier No. H00006100K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human RP9 peptide using ARM Technology.

The protein encoded by this gene can be bound and phosphorylated by the protooncogene PIM1 product, a serine/threonine protein kinase . This protein localizes in nuclear speckles containing the splicing factors, and has a role in pre-mRNA splicing. CBF1-interacting protein (CIR), a corepressor of CBF1, can also bind to this protein and effects alternative splicing. Mutations in this gene result in autosomal dominant retinitis pigmentosa-9. This gene has a pseudogene (GeneID: 441212), which is located in tandem array approximately 166kb distal to this gene. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen RP9
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description retinitis pigmentosa 9 (autosomal dominant)
Formulation In 1X PBS, pH 7.4
Gene RP9
Gene Accession No. RP9
Gene Alias PAP-1
Gene Symbols RP9
Host Species Rabbit
Immunogen A synthetic peptide of human RP9
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6100
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.