Learn More
RPA11 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA3955
Description
RPA p11 is a subunit of replication protein A, also known as Human single-stranded DNA-binding protein (HSSB). P11 and p34 are thought to form the base for the complex before p70 is added to the complex. Disruption of the p11 subunit blocks replication, due to the lack of formation of the entire HSSB complex.
PA3-955 detects RPA p11 in human cells.PA3-955 has been successfully used in Western blotting procedures. By Western blot it detects a ∽11kDa protein representing RPA p11.The PA3-955 immunogen is a synthetic peptide conjugated to the 20 amino acids of the RPA p11C-terminus.
Specifications
RPA11 | |
Polyclonal | |
HSSB; Replication Protein A | |
Synthetic peptide conjugated to the C-Terminus of RPA p11. | |
RUO | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.