Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA2 p34 Mouse anti-Human, Mouse, Clone: RPA20, 1-46, MilliporeSigma™

Catalog No. 041481MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-148-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-148-1MI Supplier MilliporeSigma Supplier No. 041481
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RPA2 p34 Antibody, clone RPA20 1-46 is a Mouse Monoclonal Antibody for detection of RPA2 p34 also known as Replication factor A protein 2 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA2 p34
Applications Western Blot
Classification Monoclonal
Clone RPA20, 1-46, 1-46
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NM_002946
Gene Symbols RPA2, REPA2, RPA32, RPA34
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human RPA2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.