Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ribosomal protein L18, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018433 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-433 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-433 Supplier Abnova Corporation Supplier No. H00006141D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human RPL18 protein.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L18E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq

Sequence: MGVDIRHNKDRKVRRKEPKSQDIYLRLLVKLYRFLARRTNSTFNQVVLKRLFMSRTNRPPLSLSRMIRKMKLPGRENKTAVVVGTITDDVRVQEVPKLKVCALRVTSRARSRILRAGGKILTFDQLALDSPKGCGTVLLSGPRKGREVYRHFGKAPGTPHSHTKPYVRSKGRKFERARGRRASRGYKN

Specifications

Antigen ribosomal protein L18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human RPL18 protein.
Formulation PBS with no preservative; pH 7.4
Gene RPL18
Gene Accession No. NM_000979.2
Gene Symbols RPL18
Host Species Rabbit
Immunogen RPL18 (NP_000970.1, 1 a.a. ∼ 188 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6141
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.