Learn More
RPL18, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RPL18 peptide using ARM Technology.
Supplier: Abnova Corporation H00006141K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L18E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RPL18 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RPL18 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
1mg | |
ribosomal protein L18 | |
RPL18 | |
RPL18 | |
A synthetic peptide of human RPL18 | |
RUO | |
6141 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.