Learn More
ribosomal protein S6 kinase, 90kDa, polypeptide 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RPS6KA3 protein.
Supplier: Abnova Corporation H00006197D01P
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in this gene have been associated with Coffin-Lowry syndrome (CLS). [provided by RefSeq
Sequence: MPLAQLADPWQKMAVESPSDSAENGQQIMDEPMGEEEINPQTEEVSIKEIAITHHVKEGHEKADPSQFELLKVLGQGSFGKVFLVKKILGSDARQLYAMKVLKKATLKVRDRVRTKMERDILVEVNHPFIVKLHYAFQTEGKLYLILDFLRGGDLFTRLSKEVMFTEEDVKFYLAELALALDHLHSLGIIYRDLKPENILLDEEGHIKLTDFGLSKESIDHEKKAYSFCGTVEYMAPEVVNRRGHTQSADWWSFGVLMFEMLTGTLPFQGKDRKETMTMILKAKLGMPQFLSPEAQSLLRMLFKRNPANRLGAGPDGVEEIKRHSFFSTIDWNKLYRREIHPPFKPATGRPEDTFYFDPEFTAKTPKDSPGIPPSANAHQLFRGFSFVAITSDDESQAMQTVGVHSIVQQLHRNSIQFTDGYEVKEDIGVGSYSVCKRCIHKATNMEFAVKIIDKSKRDPTEEIEILLRYGQHPNIITLKDVYDDGKYVYVVTELMKGGELLDKILRQKFFSEREASAVLFTITKTVEYLHAQGVVHRDLKPSNILYVDESGNPESIRICDFGFAKQLRAENGLLMTPCYTANFVAPEVLKRQGYDAACDIWSLGVLLYTMLTGYTPFANGPDDTPEEILARIGSGKFSLSGGYWNSVSDTAKDLVSKMLHVDPHQRLTAALVLRHPWIVHWDQLPQYQLNRQDAPHLVKGAMAATYSALNRNQSPVLEPVGRSTLAQRRGIKKITSTALSpecifications
ribosomal protein S6 kinase, 90kDa, polypeptide 3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human RPS6KA3 protein. | |
RPS6KA3 | |
CLS/HU-3/ISPK-1/MAPKAPK1B/MRX19/RSK/RSK2/S6K-alpha3/p90-RSK2/pp90RSK2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
Antibody |
In situ PLA, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC096303 | |
RPS6KA3 | |
RPS6KA3 (AAH96303.1, 1 a.a. ∼ 740 a.a) full-length human protein. | |
100 μg | |
Kinases | |
6197 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.