Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ribosomal protein SA, Mouse, Clone: 8G4, Abnova™

Catalog No. 89001465 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-465 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-465 Supplier Abnova Corporation Supplier No. H00003921M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RPSA.

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Many of the effects of laminin are mediated through interactions with cell surface receptors. These receptors include members of the integrin family, as well as nonintegrin laminin-binding proteins. This gene encodes a high-affinity, nonintegrin family, laminin receptor 1. This receptor has been variously called 67kDa laminin receptor, 37kDa laminin receptor precursor (37LRP) and p40 ribosome-associated protein. The amino acid sequence of laminin receptor 1 is highly conserved through evolution, suggesting a key biological function. It has been observed that the level of the laminin receptor transcript is higher in colon carcinoma tissue and lung cancer cell line than their normal counterparts. Also, there is a correlation between the upregulation of this polypeptide in cancer cells and their invasive and metastatic phenotype. Multiple copies of this gene exist, however, most of them are pseudogenes thought to have arisen from retropositional events. Two alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: EVMPDLYFYRDPEEIEKEEQAAAEKAVTKEEFQGEWTAPAPEFTATQPEVADWSEGVQVPSVPIQQFPTEDWSAQPATEDWSAAPTAQATEWVGATTDWS

Specifications

Antigen ribosomal protein SA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RPSA.
Formulation ascites with no preservative
Gene RPSA
Gene Accession No. NM_002295
Gene Alias 37LRP/67LR/LAMBR/LAMR1/LRP/p40
Gene Symbols RPSA
Host Species Mouse
Immunogen RPSA (NP_002286, 196 a.a. ∼ 295 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3921
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.