Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRN3 RNA polymerase I transcription factor homolog (S. cerevisiae), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-098 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-098 Supplier Abnova Corporation Supplier No. H00054700B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RRN3 protein.

Sequence: MRALENDFFNSPPRKTVRFGGTVTEVLLKYKKGETNDFELLKNQLLDPDIKDDQIINWLLEFRSSVMYLTKDFEQLISIILRLPWLNRSQTVVEEYLAFLGNLVSA

Specifications

Antigen RRN3 RNA polymerase I transcription factor homolog (S. cerevisiae)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RRN3 protein.
Formulation PBS with no preservative; pH 7.4
Gene RRN3
Gene Accession No. ENST00000219758
Gene Alias DKFZp566E104/MGC104238/TIFIA
Gene Symbols RRN3
Host Species Mouse
Immunogen RRN3 (ENSP00000219758, 1 a.a. ∼ 106 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 54700
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.