Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV Mouse anti-Human, Clone: 130-12H, MilliporeSigma™

Catalog No. MAB85835MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB85835MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB85835MI Supplier MilliporeSigma Supplier No. MAB85835
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RSV Antibody, nucleoprotein, clone 130-12H is an antibody against Respiratory Syncytial Virus for use in ELISA and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 130-12H
Conjugate Unconjugated
Formulation Purified. Liquid 0.02M PB, 0.25M NaCl, Ph 7.6 0.1% sodium azide.
Host Species Mouse
Immunogen A2
Quantity 500 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.