Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV Mouse anti-Human, Clone: 133-1H, MilliporeSigma™

Catalog No. MAB8581MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8581MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8581MI Supplier MilliporeSigma Supplier No. MAB8581
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H is validated for use in ELISA, immunofluorescence for the detection of Respiratory Syncytial Virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSV
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 133-1H
Conjugate Unconjugated
Formulation Ascites fluid with 0.1% sodium azide as a preservative.
Host Species Mouse
Immunogen A2
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.