Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RSV Mouse anti-Human, Clone: 133-1H, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8262
Description
Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H is an antibody against Respiratory Syncytial Virus for use in ELISA and immunofluoresence.
Specifications
RSV | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Human | |
Purified |
ELISA, Immunofluorescence | |
133-1H | |
Purified. Liquid 0.02M PBS, 0.25M NaCl, 0.1% sodium azide | |
Respiratory Syncytial virus A2 | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction